Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LB agar, Miller Microbiologically Tested, MilliporeSigma™ Supelco™

Catalog No. p-7193419 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 g
1 kg

Contains: tryptone, yeast extract, sodium chloride and agar

TRUSTED_SUSTAINABILITY

Specifications

Description Contains: tryptone, yeast extract, sodium chloride and agar
Quantity 250 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.