Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Agar plate with 25μg/mL Kanamycin

Catalog No. aaj60487eqf
Click to view available options
Quantity:
20 Plates

 

Suitable for selective cultivation of E. coli strains that contain plasmids conferring kanamycin resistance. The bacteria can be used for DNA plasmid production or production of recombinant proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Used for growth of Escherichia coli, plasmid and other bacterial cultures
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Solid
Media Type LB Agar
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging Type Plate
Quantity 20 Plates
Height (Metric) 15 mm
Diameter (Metric) Outside 10 cm

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.