Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ LB Broth, Miller
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 kg
5 kg
Description
- For the cultivation, propagation and maintenance of E. coli , B. subtilis, S. choleraesuis, C. glutamicum and other bacteria
Specifications
Specifications
| Appearance | Fine, off-white powder |
| Form | Powder |
| Media Type | LB Miller |
| Product Type | Dehydrated Culture Media |
| Sterility | Non-sterile |
| Formulation | Contains 10 g/L sodium |
| For Use With (Application) | Cultivation, Progagation, and Maintenance of Bacteria for Molecular Biology Applications |
| Packaging Type | Plastic Container |
| Quantity | 1 kg |
For research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction