Learn More
Gibco™ LB Broth (1X)

Description
LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized and can be used for nearly all applications that require a liquid bacteria culturing step, such as protein expression, cloning, and expansion.
LB broth is one of the most referenced bacterial media for cultivating and maintaining E. coli strains. Our LB broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium.
LB broth formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.
- Tryptone provides nitrogen, vitamins, minerals, and amino acids essential for growth
- Yeast extract is a source of vitamins, particularly B-group vitamins
- Sodium chloride supplies essential electrolytes for transport and osmotic balance
Primary applications
- Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
- Resuspension of T. pyriformis cell fractions
Gibco media bottles
Each media bottle features an angled opening for simplified pipetting. The 1-L Gibco media bottle uses 39% less source material (33% less for the 500-mL bottle) than comparable media bottles on the market. The cap and bottle are made from high-density polyethylene (HDPE) and polyethylene terephthalate (PETE) and can both be recycled.
Find the media format that you need
We offer other ready-to-use media formulations, such as Terrific Broth (Cat. No. A1374301) or S.O.C Recovery Medium (Cat. No. 15544034).
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • 500 mL LB Broth (1X) Store at room temperature. |
| Format | Bottle |
| Media Type | LB Broth (Lennox) |
| Preparation Method | Ready-to-Use |
| Product Type | Liquid Medium |
| Sterility | Sterile |
| Target Organism Class | Coliforms |
| Quantity | 500 mL |
| Shipping Condition | Room Temperature |
| Species | Escherichia coli |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.