Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LbCpf1, Mouse anti-Bacteria, Clone: 2D5-6G11, Millipore Sigma™

Catalog No. MABE1824100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1824100 100 μg
MABE182425 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1824100 Supplier MilliporeSigma Supplier No. MABE1824100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LbCpf1, clone 2D5-6G11 Antibody, Cat. No. MABE1824, is a highly specific mouse monoclonal antibody that targets LbCpf1 and has been tested for use in Western Blotting.

CRISPR-associated endonuclease Cas12a (UniProt: A0A182DWE3; also known as CRISPR-associated endonuclease Cpf1; Lbcpf1) is a single crRNA-guided endonuclease of the class 2 CRISPR-Cas system that lacks tracrRNA and undergoes large conformational changes upon crRNA binding. Cpf1 has several features that can distinguish it from Cas9. Unlike Cas9 that uses dual-guide RNAs, Cpf1 is guided by a crRNA. It recognizes a T-rich PAM (T-rich protospacer adjacent motif), whereas Cas9 favors a G-rich PAM and Cpf1 generates staggered ends at its PAM-distal region, but Cas9 creates blunt ends at its PAM-proximal region. Cpf1 displays higher targeting specificity in mammalian cells when compared to Cas9. It has been shown that depending on the PAM sequences, LbCpf1 can undergo conformational changes to form altered interactions with the PAM-containing DNA duplexes, hence achieving the relaxed PAM recognition. (Ref.: Zetsche, B., et al. (2015). Cell 163(3); 759-77; Yamano, T., et al. (2017). Mol. Cell 67(4); 633-645; Gao, P., et al. (2016). Cell Res. 26(8); 901-913).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LbCpf1
Applications Western Blot
Classification Monoclonal
Clone 2D5-6G11
Conjugate Unconjugated
Dilution Western Blotting Analysis: Various dilutions from a representative lot detected LbCpf1 in lysates from HEK293 cells expressing HA-tagged LbCpf1 (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Cpf1
Host Species Mouse
Immunogen His-tagged recombinant fragment from the N-terminal region of Lachnospiracea bacteria.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 2D5-6G11 specifically detects CRISPR-associated endonuclease Cas12a (LbCpf1).
Target Species Bacteria
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.