Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LBP Monoclonal Antibody (6G3)
GREENER_CHOICE

Catalog No. 01676520
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-520 Supplier Invitrogen™ Supplier No. MA140129
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40129 detects LBP from human, bovine, dog, goat, cynomologous monkey, rabbit, rat, sheep, and swine samples. Does not react with mouse or equine samples. MA1-40129 has been successfully used in immunoprecipitation, functional assays and ELISA applications. Not useful in Immunohistochemistry or Western blot applications. ELISA: monoclonal antibody 3F9 can be used both as coating and as detector. The MA1-40129 immunogen is human natural and recombinant LBP. Endotoxin <24 EU/mg.

The LBP protein is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LBP
Applications ELISA, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone 6G3
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene LBP
Gene Accession No. P17454, P18428, Q2TBI0, Q63313
Gene Alias acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88
Gene Symbols LBP
Host Species Mouse
Immunogen Human natural and recombinant LBP.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009124, 100324197, 101055392, 101104595, 102186029, 29469, 3929, 397303, 449180, 449181, 449301, 485869, 512242
Target Species Human, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Pig, Goat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.