Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LBX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564884
Description
Immunogen sequence: ITWFQNRRAKL KRDLEEMKAD VESAKKLGPS GQMDIVALAE LEQNSEATAG GGGGCGRAKS RPGSPVLPPG APKAPGAGAL QLSPASPLTD QPASSQDCSE DEEDEEIDVD Highest antigen sequence identity to the following orthologs - mouse 93%, rat 90%.
LBX1 is the transcription factor required for the development of GABAergic interneurons in the dorsal horn of the spinal cord and migration and further development of hypaxial muscle precursor cells for limb muscles, diaphragm and hypoglossal cord.This gene and the orthologous mouse gene were found by their homology to the Drosophila lady bird early and late homeobox genes. In the mouse, this gene is a key regulator of muscle precursor cell migration and is required for the acquisition of dorsal identities of forelimb muscles.
Specifications
LBX1 | |
Polyclonal | |
Unconjugated | |
LBX1 | |
EMBL:BAE92722.1}; homeobox; HPX6; HPX-6; lady bird-like homeobox; lady bird-like homeobox 1 homolog; ladybird homeobox 1; ladybird homeobox homolog 1; ladybird homeobox homolog 1 (Drosophila); ladybird homeobox protein homolog 1; Lbx1; lbx1 {ECO:0000312; Lbx1h; LOW QUALITY PROTEIN: transcription factor LBX1; RGD1564197; Transcription factor LBX1; transcription factor similar to D. melanogaster homeodomain protein lady bird late | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52954 | |
LBX1 | |
Recombinant Human LBX1. Recombinant protein control fragment (Product #RP-105687). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction