Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LBX1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1736 Supplier Bioss Supplier No. BS11867R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LBX1 is the transcription factor required for the development of GABAergic interneurons in the dorsal horn of the spinal cord and migration and further development of hypaxial muscle precursor cells for limb muscles, diaphragm and hypoglossal cord.This gene and the orthologous mouse gene were found by their homology to the Drosophila lady bird early and late homeobox genes. In the mouse, this gene is a key regulator of muscle precursor cell migration and is required for the acquisition of dorsal identities of forelimb muscles.

Specifications

Antigen LBX1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LBX1
Gene Accession No. P52954
Gene Alias BOS_23467; EMBL:BAE92722.1}; homeobox; HPX6; HPX-6; lady bird-like homeobox; lady bird-like homeobox 1 homolog; ladybird homeobox 1; ladybird homeobox homolog 1; ladybird homeobox homolog 1 (Drosophila); Ladybird homeobox protein homolog 1; Lbx1; lbx1 {ECO:0000312; Lbx1h; LOW QUALITY PROTEIN: transcription factor LBX1; RGD1564197; Transcription factor LBX1; transcription factor similar to D. melanogaster homeodomain protein lady bird late
Gene Symbols LBX1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LBX1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10660
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.