Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LC 2
GREENER_CHOICESDP

Catalog No. 74202 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-202 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 74-202 Supplier R&D Systems™ Supplier No. 7420/2

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective KRAS PROTAC™

LC 2 is a mutant-selective PROTAC™ Degrader of KRAS, comprising a ligand for the von Hippel Lindau E3 ligase joined to the KRAS inhibitor MRTX849. LC 2 induces selective degradation of KRASG12C in cancer cell lines without inducing degradation of any other mutants (DC50 = 0.25-0.76 μM), leading to suppression of MAPK signaling and modulation of ERK signaling in both homozygous and heterozygous KRASG12C cancer cell lines. LC 2 inhibits phosphorylation of ERK in SW1573 cell lines.

Negative control LC 2 Epimer (Cat. No. 7421) also available.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-(3-(3-(-2-(((7-(8-Chloronaphthalen-1-yl)-4-(-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-m)
CAS 2502156-03-6
Quantity 2 mg
Product Line Tocris
Target Active Degraders
Molecular Formula C59H71ClFN11O7S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.