Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Antibody (2312D) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP275924SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275924SS 0.025 mg
NBP275924 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP275924SS Supplier Novus Biologicals Supplier No. NBP275924SS
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LC3A Monoclonal specifically detects LC3A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LC3A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2312D
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunohistochemistry-Paraffin 3-25 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen Partial synthetic peptide from the C-terminal of human LC3A [UniProt Q9H492]
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.025 mg
Research Discipline Autophagy, Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytoskeleton Markers, Hypoxia, Macroautophagy, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84557.0
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.