Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LC3A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119167AF594
Description
LC3A Polyclonal antibody specifically detects LC3A in Human, Mouse, Rat, Bovine, Zebrafish samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
LC3A | |
Polyclonal | |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A, Microtubule-associated protein 1 light chain 3 alpha, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
Partial recombinant protein made to an N-terminal portion of the human LC3A protein [Swiss-Prot# Q9H492]. | |
0.1 mL | |
Autophagy, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Macroautophagy, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction | |
84557 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction