Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A/B Mouse anti-Human, MilliporeSigma™

Catalog No. MABC119325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1193-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1193-25 Supplier MilliporeSigma Supplier No. MABC119325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect LC3A/B using this mouse monoclonal Anti-LC3A/B, clone 4F11.1 Antibody tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3A/B
Applications Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols MAP1LC3B,MAP1ALC3
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to amino acids 2-74 of human Autophagy-related protein LC3 B.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 4F11.1 detects Autophagy-related protein LC3 B in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.