Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LC3A/LC3B Polyclonal Antibody, DyLight™ 488
GREENER_CHOICE

Catalog No. PIPA522731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522731 Supplier Invitrogen™ Supplier No. PA522731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 100% sequence homology with xenopus, bovine and zebrafish.

Autophagy is a process of intracellular bulk degradation in which cytoplasmic components, including organelles, are sequestered within double-membrane vesicles that deliver the contents to the lysosome/vacuole for degradation. During macroautophagy, the sequestering vesicles, termed autophagosomes, fuse with the lysosome or vacuole resulting in the delivery of an inner vesicle (autophagic body) into the lumen of the degradative compartment. There are 16 proteins participating in the autophagy pathway in human. The autophagy protein LC3, a mammalian homologue of Atg8, was originally identified as microtubule-associated protein 1 light chain 3. It is a component of both the MAP1A and MAP1B microtubule-binding domains and the heavy-chain independent regulation of LC3 expression might modify MAP1 microtubule-binding activity during development. LC3 is the only known mammalian protein identified that stably associates with the autophagosome membranes. LC3-I is cytosolic and LC3-II is membrane bound and enriched in the autophagic vacuole fraction. The detection of LC3-I to LC3-II conversion is a useful and sensitive marker for distinguishing autophagy in mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3A/LC3B
Applications ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate DyLight 488
Formulation 50mM sodium borate with 0.05% sodium azide
Gene MAP1LC3B2
Gene Accession No. A6NCE7, Q62625, Q6XVN8, Q91VR7, Q9CQV6, Q9GZQ8, Q9H492
Gene Alias 1010001C15Rik; 1010001H21Rik; 4922501H04Rik; Atg8; ATG8E; ATG8F; Autophagy-related protein LC3 A; autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 A; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3a; LC3b; MAP1 light chain 3-like protein 1; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 A; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 A; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; Map1lc3; Map1lc3a; MAP1LC3B; MAP1LC3B-a; microtubule associated protein 1 light chain 3 alpha; microtubule associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 alpha; Microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; Microtubule-associated proteins 1A/1B light chain 3B; Mpl3; wu:fb60g11; zbs559; zgc:56434; zgc:77094
Gene Symbols MAP1LC3A, MAP1LC3B, MAP1LC3B2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100359928, 322425, 362245, 406466, 477201, 479619, 643246, 64862, 66734, 67443, 81631, 84557
Target Species Amphibian, Canine, Fish, Human, Mouse, Plant, Rat, Zebrafish
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.