Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LC3A Monoclonal Antibody (7G10), Cyanine7, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33309MCY7

This item is not returnable.
View return policy
Description
LC3A Monoclonal antibody specifically detects LC3A in Human, Mouse, Rat samples. It is validated for ImmunocytochemistrySpecifications
LC3A | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q6XVN8, Q91VR7, Q9H492 | |
Mouse | |
Protein A | |
RUO | |
362245, 66734, 84557 | |
-20°C or -80°C if preferred | |
Liquid |
Immunocytochemistry | |
7G10 | |
Cyanine7 | |
MAP1LC3A | |
1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3a; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; map1lc3a; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; similar to microtubule-associated protein 1 light chain 3 alpha; zgc:77094 | |
KLH conjugated synthetic peptide derived from human LC3A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction