Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LC3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523180 Supplier Invitrogen™ Supplier No. PA523180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3A Polyclonal Antibody for Western Blot, IHC (P)

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene MAP1LC3A
Gene Accession No. Q6XVN8, Q91VR7, Q9H492
Gene Alias 1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3a; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; map1lc3a; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; similar to microtubule-associated protein 1 light chain 3 alpha; zgc:77094
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human ATG8 (LC3, MAP1LC3A, APG8).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362245, 66734, 84557
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.