Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Primate, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178964UV

Catalog No. NBP178964UV


Add to cart

Description

Description

LC3A Polyclonal antibody specifically detects LC3A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

LC3A
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
This LC3I antibody specifically detects the cytosolic form of LC3 before it gets converted into LC3II during autophagy.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
MAP1LC3A
A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492]
Immunogen affinity purified
Autophagy, Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytoskeleton Markers, Hypoxia, Macroautophagy, Neuroscience, Phospho Specific, Signal Transduction
Primary
84557.0
Bovine, Human, Mouse, Porcine, Primate, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit