Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LC3B Antibody (1251A), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP246892AF350
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.Specifications
LC3B | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MAP1LC3B | |
Recombinant monoclonal LC3B Antibody (1251A) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]. | |
Protein G purified | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
81631.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
1251A | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated | |
ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
Rabbit | |
14.688 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction