Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LC3B Antibody (1251B) - BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | LC3B |
---|---|
Clone | 1251B |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.1 ug/ml, Simple Western 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 25 ug/ml, Flow (Intracellular) 1 ug/ml, Knockout Validated |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP260735SS
![]() |
Novus Biologicals
NBP260735SS |
0.025 mg |
Each for $243.50
|
|
|||||
NBP260735
![]() |
Novus Biologicals
NBP260735 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.Specifications
LC3B | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, KnockDown | |
Unconjugated | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
PBS with 0.02% Sodium Azide | |
81631 | |
Recombinant monoclonal LC3B Antibody (1251B) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1251B | |
Western Blot 0.1 ug/ml, Simple Western 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 25 ug/ml, Flow (Intracellular) 1 ug/ml, Knockout Validated | |
Monoclonal | |
Rabbit | |
Human, Mouse, Rat | |
anti-LC3B, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B monoclonal, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
MAP1LC3B | |
IgG | |
Affinity Purified | |
14.688 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title