Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3B Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002220SS Shop All R&D Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002220SS 0.025 mL
NB1002220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. NB1002220SS missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NB1002220SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3B Polyclonal specifically detects LC3B in Alligator, Avian, Bacteria, Bovine, Caenorhabditis elegans, Canine, Chicken, Chinese Hamster, Drosophila melanogaster, Golden Syrian Hamster, Guinea Pig, Hamster, Human, Invertebrate, Monkey, Mouse, Pig, Primate, Rabbit, Rat, Rhesus Macaque, Zebrafish samples. It is validated for Block/Neutralize, Chemotaxis, ChIP Assay, Electron Microscopy, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, SDS-Page, Simple Western, Western Blot.

Specifications

Antigen LC3B
Applications Blocking Assay, Chemotaxis, ChIP Assay, Electron Microscopy, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Proximity Ligation Assay, SDS-Page, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2.0 ug/mL, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 20 ug/500 ug of protein, Immunohistochemistry-Paraffin 1:200 - 1:400, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9GZQ8
Gene Alias anti-LC3B, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Gene Symbols MAP1LC3B
Host Species Rabbit
Immunogen Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]
Molecular Weight of Antigen 14.688 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81631
Test Specificity Human, rat, mouse, zebrafish, bacteria, hamster, and porcine. Bovine reactivity reported in scientific literature (PMID: 24895572) Primate reactivity reported in scientific literature (PMID: 25142602) The mouse detection has been reported to be weaker than the human. Immunogen sequence has 84% homology to Xenopus.
Target Species Alligator, Mouse, Avian, Bacteria, Bovine, Avian, C. elegans, Bovine, Canine, Chicken, Guinea Pig, Chicken, Chinese Hamster, Drosophila melanogaster, Golden Syrian Hamster, Guinea Pig, Hamster, Human, Invertebrate, Monkey, Mouse, Pig, Primate, Rabbit, Rat, Rhesus Macaque, Zebrafish
Content And Storage Store at -20°C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.