Learn More
Description
Specifications
Specifications
| Antigen | LC3B |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Immunohistochemistry (Free Floating), KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.1-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen 1:400, Immunoblotting, Electron Microscopy, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q9GZQ8 |
| Gene Alias | ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B |
| Gene Symbols | MAP1LC3B |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
