Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LC3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30328020UL
Description
LC3C Polyclonal specifically detects LC3C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LC3C | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
ATG8J, Autophagy-related protein LC3 C, Autophagy-related ubiquitin-like modifier LC3 C, LC3C, LC3-like protein 2, MAP1 light chain 3-like protein 2, MAP1 light chain 3-like protein 3, MAP1A/MAP1B LC3 C, microtubule-associated protein 1 light chain 3 gammaMAP1A/MAP1B light chain 3 C, microtubule-associated proteins 1A/1B light chain 3C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human LC3C (NP_001004343.1). | |
20 μL | |
Autophagy, Cellular Markers, Phospho Specific | |
440738 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction