Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3C, Rabbit anti-Rat, Polyclonal, Millipore Sigma™

Catalog No. ABC2075200 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC2075200 200 μL
ABC207550UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC2075200 Supplier MilliporeSigma Supplier No. ABC2075200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-LC3C, Cat. No. ABC2075, is a rabbit polyclonal antibody that detects Microtubule-associated proteins 1A/1B light chain 3C and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Microtubule-associated proteins 1A/1B light chain 3C (UniProt: Q9BXW4; also known as Autophagy-related protein LC3C, Autophagy-related ubiquitin-like modifier LC3C, MAP1 light chain 3-like protein 3, MAP1A/MAP1B light chain 3 C, MAP1A/MAP1B LC3 C, Microtubule-associated protein 1 light chain 3 gamma) is encoded by the MAP1LC3C gene (Gene ID: 440738) in human. LC3s (MAP1-LC3A, B and C) are structural proteins of autophagosomal membranes, widely used as biomarkers of autophagy. LC3A is mainly localized to perinuclear and nuclear regions, LC3B is evenly distributed throughout the cytoplasm and is also localized in the nucleolar regions, and LC3C is shown to be located in the cytoplasm and strongly in the nuclei. LC3C is synthesized with a propeptide (aa 127-147), which is removed in the mature form. LC3C is a ubiquitin-like modifier that plays a crucial role in antibacterial autophagy through the selective binding of CALCOCO2. It recruits all ATG8 family members to infecting bacteria such as S.Typhimurium. It is also reported to play a role in aggrephagy, the macroautophagic degradation of ubiquitinated and aggregated proteins. LC3C is recruited to damaged mitochondria where it mediates autophagosomal degradation of mitochondrial parts in a piecemeal fashion and is required for maintaining basal mitochondrial network homeostasis. It also mediates autophagosomal degradation of MTX1 protein and cooperates with p62 to deliver MTX1 to lysosomes. (Ref.: Koukourakis, KI et al. (2015). PLoS One DOI:10.1371/journal.pone.0137675; Le Guerroue´ et al., 2017, Mol. Cell 68(4); 786-796).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected LC3C in rat brain tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Microtubule-associated proteins 1A/1B light chain 3C;Autophagy-related protein LC3 C;Autophagy-related ubiquitin-like modifier LC3C;MAP1 light chain 3-like protein 3;MAP1A/MAP1B light chain 3 C;MAP1A/MAP1B LC3 C;Microtubule-associated protein 1 light chain 3 gamma
Gene Symbols MAP1LC3C
Host Species Rabbit
Immunogen GST-tagged full-length human recombinant Microtubule-associated proteins 1A/1B light chain 3C (LC3C).
Molecular Weight of Antigen ∽17 kDa observed; 16.85 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001004343.1
Test Specificity This rabbit polyclonal antibody specifically detects LC3C.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.