Learn More
Description
Anti-LC3C, Cat. No. ABC2075, is a rabbit polyclonal antibody that detects Microtubule-associated proteins 1A/1B light chain 3C and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
Specifications
| Antigen | LC3C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected LC3C in rat brain tissue sections. |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Gene Alias | Microtubule-associated proteins 1A/1B light chain 3C;Autophagy-related protein LC3 C;Autophagy-related ubiquitin-like modifier LC3C;MAP1 light chain 3-like protein 3;MAP1A/MAP1B light chain 3 C;MAP1A/MAP1B LC3 C;Microtubule-associated protein 1 light chain 3 gamma |
| Gene Symbols | MAP1LC3C |
| Host Species | Rabbit |
| Immunogen | GST-tagged full-length human recombinant Microtubule-associated proteins 1A/1B light chain 3C (LC3C). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.