Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100601 Supplier Invitrogen™ Supplier No. PA5100601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LCA5.

Might be involved in minus end-directed microtubule transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCA5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LCA5
Gene Accession No. Q80ST9, Q86VQ0
Gene Alias 4930431B11Rik; 5730406O13Rik; AV274874; C6orf152; LCA5; LCA5, lebercilin; Leber congenital amaurosis 5; Leber congenital amaurosis 5 (human); leber congenital amaurosis 5 protein; Leber congenital amaurosis 5 protein homolog; lebercilin; ORF64; RGD1308555
Gene Symbols LCA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human LCA5(Accession Q86VQ0), corresponding to amino acid residues F639-D689.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167691, 75782
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.