Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144746 Supplier Invitrogen™ Supplier No. PA5144746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQAPRKPSPK GLPNRKGVRV GFRSQSLNRE PLRKDTDLVT KRILSARLLK INELQNEVSE LQVKLAELLK ENKSLKRLQY RQEKALNKFE DAENEISQL Antigen sequence identity: Mouse 85%; Rat 86%.

Might be involved in minus end-directed microtubule transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCA5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCA5
Gene Accession No. Q86VQ0
Gene Alias 4930431B11Rik; 5730406O13Rik; AV274874; C6orf152; LCA5; LCA5, lebercilin; Leber congenital amaurosis 5; Leber congenital amaurosis 5 (human); leber congenital amaurosis 5 protein; Leber congenital amaurosis 5 protein homolog; lebercilin; ORF64; RGD1308555
Gene Symbols LCA5
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 62-160. Recombinant protein control fragment (Product #RP-109988).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.