Learn More
LCA5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LCA5 |
---|---|
Concentration | 0.42 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2385
![]() ![]() |
Proteintech Group Inc
193331AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
Might be involved in minus end-directed microtubule transport.Specifications
LCA5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q86VQ0 | |
167691 | |
LCA5 Fusion Protein Ag5140 | |
Primary | |
-20°C | |
LCA5 |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C6orf152, LCA5, Leber congenital amaurosis 5, Lebercilin | |
LCA5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.