Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCA5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565114 Supplier Invitrogen™ Supplier No. PA565114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILPFTSMRHQG TQKSDVPPLT TKGKKATGNI DHKEKSTEIN HEIPHCVNKL PKQEDSKRKY EDLSGEEKHL EVQILLENT Highest antigen sequence identity to the following orthologs - mouse 55%, rat 54%.

LCA5L gene ontology annotations related to this gene include intraciliary transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCA5L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCA5L
Gene Accession No. O95447
Gene Alias 4921526F01Rik; C21orf13; lca5l; LCA5L, lebercilin like; Lcca5l; Leber congenital amaurosis 5-like; leber congenital amaurosis 5-like protein; Lebercilin-like protein
Gene Symbols LCA5L
Host Species Rabbit
Immunogen Recombinant Human LCA5L. Recombinant protein control fragment (Product #RP-105874).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.