Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522965 Supplier Invitrogen™ Supplier No. PA522965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for LCAT (transient overexpression lysate), Lipoprotein fraction of whole mouse plasma.

Lecithin: cholesterol acyltransferase (LCAT) is the plasma enzyme responsible for most HDL cholesteryl esters in human plasma. In species with cholesteryl ester transfer protein (CETP) it is also responsible for a significant proportion of VLDL and LDL cholesteryl esters. LCAT, through the esterification of cholesterol, plays a role in the reverse cholesterol transport pathway by facilitating the net movement of cholesterol from peripheral tissues back to the liver for excretion. LCAT deficiency results in low levels of plasma HDL, whereas a transgenic overexpression of LCAT results in increased plasma HDL concentrations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCAT
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene LCAT
Gene Accession No. P04180, P16301
Gene Alias AI046659; D8Wsu61e; Lcat; lecithin cholesterol acyltransferase; lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; phosphatidylcholine-sterol acyltransferase; phosphatidylcholine--sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase; testicular secretory protein Li 24
Gene Symbols LCAT
Host Species Rabbit
Immunogen Carboxyl-terminal histidine-tagged mouse recombinant LCAT purified from the medium of CHO cells overexpressing mouse LCAT cDNA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16816, 3931
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.