Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCAT Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143295
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
Lecithin: cholesterol acyltransferase (LCAT) is the plasma enzyme responsible for most HDL cholesteryl esters in human plasma. In species with cholesteryl ester transfer protein (CETP) it is also responsible for a significant proportion of VLDL and LDL cholesteryl esters. LCAT, through the esterification of cholesterol, plays a role in the reverse cholesterol transport pathway by facilitating the net movement of cholesterol from peripheral tissues back to the liver for excretion. LCAT deficiency results in low levels of plasma HDL, whereas a transgenic overexpression of LCAT results in increased plasma HDL concentrations.
Specifications
| LCAT | |
| Polyclonal | |
| Unconjugated | |
| LCAT | |
| AI046659; D8Wsu61e; Lcat; lecithin cholesterol acyltransferase; lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; phosphatidylcholine-sterol acyltransferase; phosphatidylcholine--sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase; testicular secretory protein Li 24 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3931 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P04180 | |
| LCAT | |
| Peptide with sequence C-EDGDDTVATRSTE (aa 366-378). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction