Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCAT Recombinant Rabbit Monoclonal Antibody (9C2U3)
GREENER_CHOICE

Catalog No. PIMA537866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537866 Supplier Invitrogen™ Supplier No. MA537866
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse plasma, Rat plasma Immunogen sequence: MGPPGSPWQW VTLLLGLLLP PAAPFWLLNV LFPPHTTPKA ELSNHTRPVI LVPGCLGNQL EAKLDKPDVV NWMCYRKTED FFTIWLDLNM FLPLGVDCWI.

Lecithin: cholesterol acyltransferase (LCAT) is the plasma enzyme responsible for most HDL cholesteryl esters in human plasma. In species with cholesteryl ester transfer protein (CETP) it is also responsible for a significant proportion of VLDL and LDL cholesteryl esters. LCAT, through the esterification of cholesterol, plays a role in the reverse cholesterol transport pathway by facilitating the net movement of cholesterol from peripheral tissues back to the liver for excretion. LCAT deficiency results in low levels of plasma HDL, whereas a transgenic overexpression of LCAT results in increased plasma HDL concentrations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCAT
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9C2U3
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LCAT
Gene Accession No. P04180, P16301, P18424
Gene Alias AI046659; D8Wsu61e; Lcat; lecithin cholesterol acyltransferase; lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; phosphatidylcholine-sterol acyltransferase; phosphatidylcholine--sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase; testicular secretory protein Li 24
Gene Symbols LCAT
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LCAT (P04180).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16816, 24530, 3931
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.