Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCE1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144236
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144236 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144236 Supplier Invitrogen™ Supplier No. PA5144236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LCE1B Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Human LCE1B (Late cornified envelope protein 1B), also named late envelope protein 2 (LEP2), is located at human Chromosome1q21 which contains multiple LCE clusters. LCE1B gene is predicated to encode a 118-aa protein, and is currently evidenced at transcript level. Its expression was skin-specific and was readily detected in adult trunk skin, adult arm skin, fetal skin, penal skin, vulva, esophagus and tongue. LCE genes respond to environmental stimuli such as calcium levels and ultraviolet (UV) light, however, LCE groups have distinct functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCE1B
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LCE1B
Gene Accession No. Q5T7P3
Gene Alias 1110029C13Rik; late cornified envelope 1B; late cornified envelope protein 1B; Late envelope protein 2; LCE1B; LEP2; small proline rich-like (epidermal differentiation complex) 2A; small proline rich-like 5; small proline-rich-like epidermal differentiation complex protein 2A; SPRL2A; Sprrl5
Gene Symbols LCE1B
Host Species Rabbit
Immunogen Recombinant Human Late cornified envelope protein 1B protein (1-118AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353132
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.