Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCE2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523538
Description
LCE2B Polyclonal Antibody for Western Blot, Flow
This protein is skin-specific.
Specifications
LCE2B | |
Polyclonal | |
Unconjugated | |
LCE2B | |
late cornified envelope 2B; late cornified envelope protein 2B; Late envelope protein 10; LCE2B; LEP10; Skin-specific protein Xp5; small proline rich-like (epidermal differentiation complex) 1B; small proline-rich-like epidermal differentiation complex protein 1B; SPRL1B; XP5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
26239 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O14633 | |
LCE2B | |
KLH conjugated synthetic peptide between 61-90 amino acids from the C-terminal region of human LCE2B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction