Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCHN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566393 Supplier Invitrogen™ Supplier No. PA566393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FYVNVADIESL EVEVSYVACT TEKIFEEKRE LYDVYVDNQN VKTHHDHLQP LLKINSADRE KYRRLNEQRQ MLL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.

Probable guanine nucleotide exchange factor (GEF). May promote the exchange of GDP to GTP, converting inactive GDP-bound small GTPases into their active GTP-bound form (Probable). May play a role in neuritogenesis, as well as in neuronal recovery and/or restructuring in the hippocampus following transient cerebral ischemia. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCHN
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND11
Gene Accession No. A4D1U4
Gene Alias AI841796; DENN domain containing 11; DENN domain-containing protein 11; Dennd11; E330009J07Rik; KIAA1147; LCHN; mKIAA1147; predicted protein of HQ2561; PRO2561; protein LCHN; RGD1563986; RIKEN cDNA E330009J07 gene; si:ch211-238n5.4; similar to RIKEN cDNA E330009J07 gene; wu:fa55h08
Gene Symbols DENND11
Host Species Rabbit
Immunogen Recombinant Human LCHN. Recombinant protein control fragment (Product #RP-107066).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.