Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCHN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597895 Supplier Thermo Scientific Supplier No. PA597895
Add to Cart
Add to Cart

LCHN Polyclonal Antibody

LCHN Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen LCHN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DENND11
Gene Accession No. A4D1U4
Gene Alias AI841796; DENN domain containing 11; DENN domain-containing protein 11; Dennd11; E330009J07Rik; KIAA1147; LCHN; mKIAA1147; predicted protein of HQ2561; PRO2561; protein LCHN; RGD1563986; RIKEN cDNA E330009J07 gene; si:ch211-238n5.4; similar to RIKEN cDNA E330009J07 gene; wu:fa55h08
Gene Symbols DENND11
Host Species Rabbit
Immunogen Recombinant Human Protein LCHN protein (2-233AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57189
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.