Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lck Antibody (33D196), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056678FR
Description
Lck Monoclonal specifically detects Lck in Human, Mouse samples. It is validated for Western Blot.Specifications
Lck | |
Monoclonal | |
DyLight 680 | |
P06239 | |
LCK | |
This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein. | |
0.2 mL | |
Adaptive Immunity, Cell Cycle and Replication, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3932 | |
Human, Mouse | |
Purified |
Western Blot | |
33D196 | |
Western Blot | |
EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Lck (33D196) | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction