Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lck Antibody (LCK-01), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500336C
Description
Lck Monoclonal specifically detects Lck in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Lck | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
LCK | |
Peptide corresponding to amino acids 22-36 in the sequence of human Lck. | |
0.1 mL | |
Primary | |
The antibody LCK-01 recognizes defined epitope (aa 22-36) of Lck, a 56 kDa Src-family protein tyrosine kinase. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LCK-01 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3932.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction