Learn More
Promega LCK Kinase Enzyme System, 1mg
Catalog No. PRV2692 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2692 1 mg
1 options
Catalog No. PRV2692 Supplier Promega Supplier No. V2692
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human LCK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LCK (p56lck) is a member of the src family of nonreceptor tyrosine kinases.
  • Easily Screen and Profile LCK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C