Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCK, Rabbit anti-Human, Clone: A3, Abnova™

Catalog No. 89335757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-757 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-757 Supplier Abnova Corporation Supplier No. MAB18923
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen LCK
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone A3
Conjugate Unconjugated
Dilution Flow Cytometry Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).
Gene lymphocyte-specific protein tyrosine kinase
Gene Alias YT16/p56lck/pp58lck
Gene Symbols LCK
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y505 of human LCK.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3932
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.