Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCLAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556886 Supplier Invitrogen™ Supplier No. PA556886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHPRTTGFTF VVDRLREGKN LDAVHDITVA YPHNIPQSEK HLLQGDFPRE IHFHVHRYPI DTLPTSKEDL QLWCHKR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

Exhibits acyl-CoA:lysocardiolipin acyltransferase (ALCAT) activity; catalyzes the reacylation of lyso-cardiolipin to cardiolipin (CL), a key step in CL remodeling. Recognizes both monolysocardiolipin and dilysocardiolipin as substrates with a preference for linoleoyl-CoA and oleoyl-CoA as acyl donors. Also exhibits 1-acyl-sn-glycerol-3-phosphate acyltransferase activity (AGPAT) activity; converts 1-acyl-sn-glycerol-3- phosphate (lysophosphatidic acid or LPA) into 1,2-diacyl-sn-glycerol-3- phosphate (phosphatidic acid or PA) by incorporating an acyl moiety at the sn-2 position of the glycerol backbone (PubMed:16620771). Possesses both lysophosphatidylinositol acyltransferase (LPIAT) and lysophosphatidylglycerol acyltransferase (LPGAT) activities (PubMed:19075029). Required for establishment of the hematopoietic and endothelial lineages. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCLAT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCLAT1
Gene Accession No. Q6UWP7
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-AGP acyltransferase 8; 1-AGPAT 8; 1AGPAT8; Acyl-CoA:lysocardiolipin acyltransferase 1; Agpat8; AI181996; Alcat1; Gm91; HSRG1849; LCLAT1; Lycat; lysocardiolipin acyltransferase 1; RGD1565906; UNQ1849; UNQ1849/PRO3579
Gene Symbols LCLAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCLAT1. Recombinant protein control fragment (Product #RP-95708).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.