Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCLAT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989852
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9852 Supplier Bioss Supplier No. BS18190R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acyl-CoA:lysocardiolipin acyltransferase. Possesses both lysophosphatidylinositol acyltransferase (LPIAT) and lysophosphatidylglycerol acyltransferase (LPGAT) activities. Recognizes both monolysocardiolipin and dilysocardiolipin as substrates with a preference for linoleoyl-CoA and oleoyl-CoA as acyl donors. Acts as a remodeling enzyme for cardiolipin, a major membrane polyglycerophospholipid. Converts lysophosphatidic acid (LPA) into phosphatidic acid (PA) with a relatively low activity. Required for establishment of the hematopoietic and endothelial lineages.

Specifications

Antigen LCLAT1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene LCLAT1
Gene Accession No. Q6UWP7
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-AGP acyltransferase 8; 1-AGPAT 8; 1AGPAT8; Acyl-CoA:lysocardiolipin acyltransferase 1; AGPAT8; AI181996; ALCAT1; Gm91; HSRG1849; LCLAT1; Lycat; Lysocardiolipin acyltransferase 1; UNQ1849; UNQ1849/PRO3579
Gene Symbols LCLAT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LCLAT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253558
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.