Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCMT1 Antibody (OTI2C9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271927H
Description
LCMT1 Monoclonal specifically detects LCMT1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LCMT1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LCMT1 | |
Full length recombinant protein of human LCMT1 (NP_057393) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2C9 | |
Western Blot, Immunohistochemistry | |
CGI-68, EC 2.1.1.-, LCMT, leucine carboxyl methyltransferase 1, PPMT1, protein phosphatase methyltransferase 1, Protein-leucine O-methyltransferase | |
Mouse | |
Affinity Purified | |
Cancer | |
51451.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction