Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCMT1 Antibody (OTI2C9), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271927PE
Description
LCMT1 Monoclonal antibody specifically detects LCMT1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
LCMT1 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI2C9 | |
Western Blot, Immunohistochemistry | |
CGI-68, EC 2.1.1.-, LCMT, leucine carboxyl methyltransferase 1, PPMT1, protein phosphatase methyltransferase 1, Protein-leucine O-methyltransferase | |
Full length human recombinant protein of human LCMT1(NP_057393) produced in HEK293T cell. | |
0.1 mL | |
Cancer | |
51451 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction