Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCMT1 Monoclonal Antibody (4A4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518059
Description
.
Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.
Specifications
LCMT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q6P4Z6 | |
LCMT1 | |
6xHis mouse PPM1. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
4A4 | |
Unconjugated | |
LCMT1 | |
[phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase | |
Mouse | |
Protein G | |
RUO | |
30949, 361643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction