Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCMT1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503181

Description
Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.Specifications
LCMT1 | |
Monoclonal | |
0.32 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UIC8 | |
LCMT1 | |
Full length human recombit protein of human LCMT1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI 5D5 | |
Unconjugated | |
LCMT1 | |
CGI-68, LCMT, PPMT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51451 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction