Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCMT1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503181S


Description
LCMT1 Monoclonal antibody specifically detects LCMT1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
LCMT1 | |
Monoclonal | |
0.32 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P22059 | |
Mouse | |
Affinity chromatography | |
RUO | |
5007 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI 5D5 | |
Unconjugated | |
Osbp | |
[phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase | |
Full length human recombinant protein of human OSBP produced in HEK293 cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction