Learn More
LCMT1 Mouse anti-Human, Clone: OTI1E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503123

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.Specifications
LCMT1 | |
Monoclonal | |
Unconjugated | |
LCMT1 | |
CGI-68, LCMT, PPMT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51451 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UIC8 | |
LCMT1 | |
Full length human recombit protein of human LCMT1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.