Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCMT1 Mouse anti-Human, Mouse, Rat, Clone: OTI2C9, liquid, TrueMAB™
SDP

Catalog No. 501674092
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4092 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-4092 Supplier OriGene Supplier No. TA503125S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.

Specifications

Antigen LCMT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C9
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LCMT1
Gene Accession No. Q6P4Z6, Q9UIC8
Gene Alias CGI-68, LCMT, PPMT1
Gene Symbols Lcmt1
Host Species Mouse
Immunogen Full length human recombit protein of human LCMT1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30949, 361643, 51451
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.