Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559622 Supplier Invitrogen™ Supplier No. PA559622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLSNGWETA SAVDMMELYN RLPRAEVSRI ESLEFLDEME LLEQLMRHYC LCWATKGGNE LGLKE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCMT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LCMT1
Gene Accession No. Q6P4Z6, Q9UIC8
Gene Alias [phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase
Gene Symbols LCMT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LCMT1. Recombinant protein control fragment (Product #RP-98805).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30949, 361643, 51451
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.