Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCMT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219340
Description
LCMT2 Polyclonal specifically detects LCMT2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LCMT2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
EC 2.1.1.-, FLJ51321, KIAA0547tRNA-Wybutosine-synthesizing protein 4, leucine carboxyl methyltransferase 2, MGC9534, p21WAF1/CIP1 promoter-interacting protein, PPM2, tRNA wybutosine-synthesizing protein 4 homolog, tRNA yW-synthesizing protein 4 homolog, TYW4tRNA-yW synthesizing protein 4 | |
Rabbit | |
76 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.03mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O60294 | |
LCMT2 | |
Recombinant protein encompassing a sequence within the center region of human LCMT2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
9836 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction