Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LCMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532055 Supplier Invitrogen™ Supplier No. PA532055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TYW4 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW4 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCMT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene LCMT2
Gene Accession No. O60294, Q8BYR1
Gene Alias D330024M17; KIAA0547; LCMT2; Leucine carboxyl methyltransferase 2; p21WAF1/CIP1 promoter-interacting protein; PPM2; tRNA wybutosine-synthesizing protein 4; tRNA wybutosine-synthesizing protein 4 homolog; tRNA yW-synthesizing protein 4; tRNA yW-synthesizing protein 4 homolog; tRNA(Phe) (7-(3-amino-3-(methoxycarbonyl)propyl)wyosine(37)-N)-methoxycarbonyltransferase; tRNA(Phe) (7-(3-amino-3-carboxypropyl)wyosine(37)-O)-methyltransferase; tRNA-Wybutosine-synthesizing protein 4; tRNA-yW synthesizing protein 4; TYW4
Gene Symbols LCMT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LCMT2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329504, 9836
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.